Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037810.2 | NP_001032899.1 | 2 | 558 | ||
NM_009259.5 | NP_033285.1 | 2 | 558 | |||
GenBank mRNA | AK041480.1 | - | 2 | 550 | ||
AK041712.1 | - | 2 | 550 | |||
AK077764.1 | - | 2 | 549 | |||
AK134098.1 | - | 2 | 553 | |||
BC100738.1 | - | 1 | 450 | 40045521 | ||
BC100739.1 | - | 1 | 450 | 40045520 | ||
BC100740.1 | - | 1 | 450 | 40045519 | ||
BC100741.1 | - | 1 | 450 | 40045517 | ||
S70677.1 | - | 1 | 450 |