Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sptbn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009260.2 | NP_033286.2 | 10 | 1626 | ||
NM_175836.2 | NP_787030.2 | 10 | 1286 | |||
GenBank mRNA | AF017112.1 | - | 10 | 1533 | ||
AK157727.1 | - | 10 | 1616 | |||
AK220456.1 | - | 11 | 1488 | |||
M74773.1 | - | 10 | 1606 |