Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sptbn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009260.2 | NP_033286.2 | 19 | 4600 | ||
NM_175836.2 | NP_787030.2 | 19 | 4260 | |||
GenBank mRNA | AF017112.1 | - | 19 | 4507 | ||
AK220456.1 | - | 20 | 4462 | |||
U73171.1 | - | 1 | 40 |