Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ING2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291959.1 | NP_001278888.1 | 2 | 748 | ||
NM_001564.3 | NP_001555.1 | 2 | 829 | |||
XM_011531927.2 | XP_011530229.1 | 2 | 674 | |||
GenBank mRNA | AB012853.1 | - | 2 | 774 | ||
AB196793.1 | - | 2 | 623 | |||
AF053537.1 | - | 2 | 750 | |||
AJ006851.1 | - | 2 | 683 | |||
AK294310.1 | - | 2 | 770 | |||
AK314000.1 | - | 2 | 830 | |||
BC030128.2 | - | 2 | 809 | 3941655 |