Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ING2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001564.3 | NP_001555.1 | 1, 2 | 310 | ||
GenBank mRNA | AB012853.1 | - | 1, 2 | 255 | ||
AF053537.1 | - | 1, 2 | 231 | |||
AJ006851.1 | - | 1, 2 | 164 | |||
AK314000.1 | - | 1, 2 | 311 | |||
BC030128.2 | - | 1, 2 | 290 | 3941655 |