Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011467.2 | NP_035597.2 | 1 | 311 | ||
GenBank mRNA | AK004941.1 | - | 1 | 303 | ||
AK154720.1 | - | 315 | ||||
BC010400.1 | - | 1 | 308 | 4219245 | ||
BC054423.1 | - | 1 | 295 | 4985039 | ||
BY113096.1 | - | 1 | 312 | |||
CK620382.1 | - | 1 | 279 | |||
S77493.1 | - | 1 | 313 |