Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ING2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291959.1 | NP_001278888.1 | 2 | 798 | ||
NM_001564.3 | NP_001555.1 | 2 | 879 | |||
XM_011531927.2 | XP_011530229.1 | 2 | 724 | |||
GenBank mRNA | AB012853.1 | - | 2 | 824 | ||
AB196793.1 | - | 2 | 673 | |||
AF053537.1 | - | 2 | 800 | |||
AJ006851.1 | - | 2 | 733 | |||
AK294310.1 | - | 2 | 820 | |||
AK314000.1 | - | 2 | 880 | |||
BC030128.2 | - | 2 | 859 | 3941655 |