Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INHBA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002192.2 | NP_002183.1 | 2 | 546 | ||
XM_017012174.1 | XP_016867663.1 | 4 | 2438 | |||
XM_017012175.1 | XP_016867664.1 | 2 | 2122 | |||
XM_017012176.1 | XP_016867665.1 | 4 | 2306 | |||
GenBank mRNA | AF218018.1 | - | 2 | 505 | ||
AK222742.1 | - | 1 | 375 | |||
AK290584.1 | - | 2 | 509 | |||
AU137531.1 | - | 2 | 546 | |||
BC007858.2 | - | 2 | 516 | 4299832 | ||
J03634.1 | - | 1 | 384 | |||
M13436.1 | - | 1 | 535 |