Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srp14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009273.4 | NP_033299.1 | 4, 5 | 334 | ||
GenBank mRNA | AK019310.1 | - | 4, 5 | 317 | ||
AK156845.1 | - | 274 | ||||
AK167821.1 | - | 4, 5 | 283 | |||
AK168144.1 | - | 4, 5 | 302 | |||
BB848168.1 | - | 4, 5 | 351 | |||
BC021537.1 | - | 4, 5 | 310 | 5353575 | ||
M29264.1 | - | 4, 5 | 259 |