Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srpk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009274.2 | NP_033300.2 | 10 | 1490 | ||
GenBank mRNA | AB006036.1 | - | 10 | 1280 | ||
AK136861.1 | - | 10 | 1456 | |||
BC020178.1 | - | 10 | 1387 | 4507346 | ||
BC062941.1 | - | 10 | 1409 |