Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srprb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009275.4 | NP_033301.1 | 1 | 68 | ||
GenBank mRNA | AK004854.1 | - | 1 | 101 | ||
AK007859.1 | - | 1 | 74 | |||
AK008383.1 | - | 1 | 111 | |||
AK015469.1 | - | 1 | 133 | |||
AK030437.1 | - | 1 | 69 | |||
AK075583.1 | - | 1 | 71 | |||
AK160600.1 | - | 1 | 134 | |||
AK167039.1 | - | 1 | 74 | |||
BC003798.1 | - | 1 | 89 | 3490766 | ||
U17343.1 | - | 1 | 82 |