Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trove2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013835.2 | NP_038863.1 | 8 | 1605 | ||
GenBank mRNA | AK076167.1 | - | 8 | 1583 | ||
AK167379.1 | - | 8 | 1546 | |||
AK167555.1 | - | 8 | 1546 | |||
AK167645.1 | - | 8 | 1546 | |||
BC040368.1 | - | 6 | 2279 | |||
BC051974.1 | - | 7 | 1482 | 5697399 | ||
BC052380.1 | - | 7 | 1482 | 5697399 | ||
L81154.1 | - | 7 | 1453 | |||
U66843.1 | - | 8 | 1532 |