Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp143 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009281.3 | NP_033307.2 | 11 | 1093 | ||
GenBank mRNA | AF011758.2 | - | 1042 | |||
AK029078.1 | - | 11 | 1085 | |||
AK029926.1 | - | 11 | 1358 | |||
BC037658.1 | - | 10 | 1003 | 5356405 |