Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019963.1 | NP_064347.1 | 3, 4 | 435 | ||
GenBank mRNA | AF206162.1 | - | 3, 4 | 435 | ||
AK149838.1 | - | 3, 4 | 432 | |||
AK149924.1 | - | 3, 4 | 448 | |||
AK150070.1 | - | 3, 4 | 447 | |||
AK153405.1 | - | 1, 2 | 73 |