Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stat4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK037598.1 | - | 2 | 227 | ||
AK132675.1 | - | 4 | 414 | |||
AK157083.1 | - | 3 | 288 | |||
BC052907.1 | - | 3 | 293 | |||
BC098499.1 | - | 3 | 281 | 30074608 | ||
BY750146.1 | - | 3 | 279 | |||
U06923.1 | - | 3 | 284 | |||
U09351.1 | - | 3 | 738 |