Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stat6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009284.2 | NP_033310.2 | 16, 17 | 2171 | ||
GenBank mRNA | AF481808.1 | - | 15, 16 | 1886 | ||
AF481809.1 | - | 15, 16 | 1886 | |||
AK149563.1 | - | 16, 17 | 2131 | |||
AK163053.1 | - | 16, 17 | 2172 | |||
AK206252.1 | - | 11 | ||||
BC029318.1 | - | 3, 4 | 360 | |||
BC127054.1 | - | 16, 17 | 1961 | 40109712 |