Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sult2a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009286.2 | NP_033312.2 | 1 | 168 | ||
GenBank mRNA | AK149424.1 | - | 1 | 169 | ||
BC125365.1 | - | 141 | 40129880 | |||
BC125367.1 | - | 141 | 40129882 | |||
L27121.1 | - | 141 |
Gene Symbol: Sult2a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111296.2 | NP_001104766.1 | 1 | 169 | ||
GenBank mRNA | BC145167.1 | - | 1 | 141 | ||
L02335.1 | - | 1 | 142 |