Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016737.2 | NP_058017.1 | 6 | 926 | ||
GenBank mRNA | AK075988.1 | - | 6 | 864 | ||
AK088494.1 | - | 6 | 868 | |||
AK149493.1 | - | 6 | 842 | |||
AK161645.1 | - | 6 | 845 | |||
AK167273.1 | - | 6 | 860 | |||
AK168181.1 | - | 6 | 842 | |||
AK189522.1 | - | 2 | 271 | |||
BC003794.1 | - | 6 | 835 | 3590457 | ||
U27830.1 | - | 6 | 843 |