Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016737.2 | NP_058017.1 | 7 | 959 | ||
GenBank mRNA | AK088494.1 | - | 7 | 901 | ||
AK149493.1 | - | 7 | 875 | |||
AK161645.1 | - | 7 | 878 | |||
AK167273.1 | - | 7 | 893 | |||
AK168181.1 | - | 7 | 875 | |||
AK189522.1 | - | 3 | 304 | |||
BC003794.1 | - | 7 | 868 | 3590457 | ||
U27830.1 | - | 7 | 876 |