Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stk16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277992.1 | NP_001264921.1 | 5 | 751 | ||
NM_011494.5 | NP_035624.3 | 5 | 779 | |||
GenBank mRNA | AA183397.1 | - | 3 | 248 | ||
AB017197.1 | - | 5 | 727 | |||
AB022179.1 | - | 5 | 709 | |||
AF062076.1 | - | 4 | 466 | |||
AF089869.1 | - | 5 | 756 | |||
AJ005790.1 | - | 5 | 710 | |||
AK030404.1 | - | 5 | 754 | |||
AK032162.1 | - | 5 | 770 | |||
AK149539.1 | - | 5 | 752 | |||
AK156554.1 | - | 4 | 834 | |||
AK212366.1 | - | 2 | 87 | |||
BC005703.1 | - | 5 | 738 | 3598470 | ||
BC028999.1 | - | 5 | 738 | 5250877 |