Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stx4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009294.3 | NP_033320.1 | 8 | 899 | ||
GenBank mRNA | AK145909.1 | - | 8 | 918 | ||
AK161971.1 | - | 8 | 892 | |||
BC005791.1 | - | 8 | 739 | 3603665 | ||
BC052023.1 | - | 8 | 835 | 5711022 | ||
U76832.1 | - | 8 | 668 |