Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stxbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011503.4 | NP_035633.1 | 5 | 296 | ||
GenBank mRNA | AF263345.1 | - | 5 | 303 | ||
AK088364.1 | - | 5 | 318 | |||
AK133419.1 | - | 5 | 297 | |||
AK182048.1 | - | 1 | 23 | |||
BC003477.1 | - | 5 | 286 | |||
BC008102.1 | - | 5 | 306 | 3966319 | ||
D42068.1 | - | 5 | 288 | |||
U19520.1 | - | 5 | 310 |