Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sucla2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011506.3 | NP_035636.1 | 9 | 1143 | ||
GenBank mRNA | AF058955.1 | - | 8 | 1010 | ||
AK003867.1 | - | 1 | 28 | |||
AK005273.1 | - | 9 | 1126 | |||
AK081965.1 | - | 9 | 1164 | |||
AK088801.1 | - | 9 | 1144 | |||
AK132762.1 | - | 9 | 1133 | |||
AK150594.1 | - | 9 | 1143 | |||
AK152783.1 | - | 9 | 1143 | |||
AK153198.1 | - | 9 | 1143 | |||
AK160130.1 | - | 9 | 1144 | |||
AK160652.1 | - | 9 | 1132 | |||
AK215074.1 | - | 3 | 259 | |||
BC038497.1 | - | 3 | 311 | |||
BC052774.1 | - | 6 | 675 | |||
BC056353.1 | - | 9 | 1134 | 6808360 | ||
BC057605.1 | - | 9 | 1134 | 6808360 |