Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sucla2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011506.3 | NP_035636.1 | 2 | 270 | ||
GenBank mRNA | AF058955.1 | - | 1 | 137 | ||
AK005273.1 | - | 2 | 252 | |||
AK081965.1 | - | 2 | 291 | |||
AK088801.1 | - | 2 | 271 | |||
AK132762.1 | - | 2 | 260 | |||
AK150594.1 | - | 2 | 270 | |||
AK152783.1 | - | 2 | 270 | |||
AK153198.1 | - | 2 | 270 | |||
AK160130.1 | - | 2 | 271 | |||
AK160652.1 | - | 2 | 259 | |||
BC056353.1 | - | 2 | 261 | 6808360 | ||
BC057605.1 | - | 2 | 261 | 6808360 |