Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Suclg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011507.3 | NP_035637.2 | 8 | 958 | ||
GenBank mRNA | AF058956.1 | - | 7 | 800 | ||
AK045176.1 | - | 8 | 911 | |||
AK156263.1 | - | 8 | 913 | |||
AK167136.1 | - | 8 | 913 | |||
AK167339.1 | - | 8 | 1197 | |||
AK172561.1 | - | 8 | 913 | |||
AK172633.1 | - | 8 | 913 | |||
AK184241.1 | - | 3 | 257 | |||
BC043312.1 | - | 6 | 653 | |||
BC054425.1 | - | 7 | 711 | |||
BC080781.1 | - | 8 | 913 | 30470296 | ||
EU234009.1 | - | 8 | 865 |