Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INPP5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297434.1 | NP_001284363.1 | 14 | 1864 | ||
NM_005540.2 | NP_005531.2 | 20 | 2371 | |||
XM_006710621.3 | XP_006710684.2 | 16 | 2134 | |||
XM_011541392.2 | XP_011539694.1 | 14 | 1810 | |||
XM_011541393.2 | XP_011539695.1 | 14 | 1799 | |||
XM_017001206.1 | XP_016856695.1 | 20 | 2408 | |||
XM_017001207.1 | XP_016856696.1 | 17 | 2111 | |||
XM_017001208.1 | XP_016856697.1 | 17 | 2240 | |||
XM_017001209.1 | XP_016856698.1 | 14 | 1790 | |||
GenBank mRNA | AK022846.1 | - | 14 | 1822 | ||
BC042529.2 | - | 13 | 1746 | |||
M74161.1 | - | 20 | 2365 |