Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Supt6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009297.2 | NP_033323.2 | 11 | 1457 | ||
GenBank mRNA | AK170533.1 | - | 11 | 1456 | ||
BC072657.1 | - | 11 | 1377 | 30536338 | ||
U40375.1 | - | 11 | 1455 |