Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcc9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044720.1 | NP_001038185.1 | 17 | 2283 | ||
NM_011511.2 | NP_035641.1 | 17 | 2283 | |||
NM_021041.2 | NP_066378.1 | 17 | 2283 | |||
NM_021042.2 | NP_066379.2 | 16 | 2178 | |||
GenBank mRNA | AF003531.1 | - | 16 | 2178 | ||
D86037.2 | - | 17 | 2227 | |||
D86038.2 | - | 17 | 2227 | |||
U97066.1 | - | 17 | 2283 |