Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Surf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009298.3 | NP_033324.1 | 2 | 282 | ||
GenBank mRNA | AK077989.1 | - | 2 | 284 | ||
AK132194.1 | - | 2 | 318 | |||
BC037634.1 | - | 2 | 329 | 4952465 | ||
X92842.1 | - | 2 | 265 |