Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sycp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011517.2 | NP_035647.2 | 1, 2 | 127 | ||
GenBank mRNA | AK131949.1 | - | 1, 2 | 51 | ||
BC048483.1 | - | 1, 2 | 50 | |||
BC089540.1 | - | 1, 2 | 50 | |||
BC099552.1 | - | 1, 2 | 50 | |||
BC132079.1 | - | 1, 2 | 128 | 40130356 | ||
BC138509.1 | - | 1, 2 | 128 | 8861529 | ||
Y08485.1 | - | 1, 2 | 136 |