Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198977.1 | NP_001185906.1 | 4, 5 | 930 | ||
NM_011518.2 | NP_035648.2 | 4, 5 | 934 | |||
GenBank mRNA | AK036736.1 | - | 4, 5 | 858 | ||
AK049207.1 | - | 5, 6 | 1028 | |||
AK136363.1 | - | 4, 5 | 933 | |||
AK143564.1 | - | 4, 5 | 963 | |||
BC065121.1 | - | 4, 5 | 866 | 30533300 | ||
U25685.1 | - | 3, 4 | 718 | |||
U36776.1 | - | 4, 5 | 937 | |||
Z49877.1 | - | 4, 5 | 1195 |
Gene Symbol: Slc4a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK171662.1 | - | 3287 | |||
AK171845.1 | - | 3287 |