Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INSIG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005542.4 | NP_005533.2 | 2 | 542 | ||
NM_198337.2 | NP_938151.1 | 2 | 542 | |||
XM_005249542.1 | XP_005249599.1 | 2 | 542 | |||
XM_005249543.1 | XP_005249600.1 | 2 | 557 | |||
XM_017012177.1 | XP_016867666.1 | 2 | 557 | |||
GenBank mRNA | AK291675.1 | - | 2 | 414 | ||
AY112745.1 | - | 1 | 331 | |||
BC001880.1 | - | 2 | 480 | 3546928 | ||
BI550548.1 | - | 2 | 493 | |||
BI918342.1 | - | 2 | 410 | |||
BQ059075.1 | - | 1 | 75 | |||
BT007227.1 | - | 1 | 331 | |||
DA738378.1 | - | 2 | 542 | |||
DN996424.1 | - | 2 | 435 |