Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syngr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009304.2 | NP_033330.1 | 4 | 952 | ||
GenBank mRNA | AF151985.1 | - | 4 | 949 | ||
AF364049.1 | - | 4 | 939 | |||
AJ002307.1 | - | 4 | 925 | |||
AK150234.1 | - | 4 | 952 | |||
AK157343.1 | - | 4 | 925 | |||
AK170681.1 | - | 4 | 952 | |||
AK208001.1 | - | 1 | 42 | |||
BC004829.1 | - | 4 | 952 | 2646657 |