Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Synj2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113351.1 | NP_001106822.1 | 22 | 3359 | ||
NM_001113352.2 | NP_001106823.1 | 23 | 3448 | |||
NM_001113353.2 | NP_001106824.1 | 24 | 3583 | |||
NM_001290698.1 | NP_001277627.1 | 17 | 2476 | |||
NM_011523.2 | NP_035653.2 | 23 | 3494 | |||
GenBank mRNA | AF041857.1 | - | 24 | 3584 | ||
AF041858.1 | - | 12 | 1711 | |||
AF041859.1 | - | 17 | 2375 | |||
AF041860.1 | - | 9 | 1291 | |||
AF041861.1 | - | 7 | 871 | |||
AF041862.1 | - | 23 | 3439 | |||
AK019694.1 | - | 17 | 2477 | |||
AK029755.1 | - | 20 | 2866 | |||
AK038038.1 | - | 20 | 2771 | |||
AK129122.1 | - | 21 | 2876 | |||
AK142919.1 | - | 23 | 3231 | |||
AK148053.1 | - | 24 | 3321 | |||
AK161467.1 | - | 17 | 2460 | |||
AK162329.1 | - | 6 | 733 | |||
BC023713.1 | - | 10 | 1395 | |||
BC058749.1 | - | 23 | 3325 | 6390688 | ||
BC060214.1 | - | 22 | 3205 | 6838657 |