Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tacc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040435.2 | NP_001035525.1 | 12, 13 | 2183 | ||
GenBank mRNA | AF093542.1 | - | 13, 14 | 1417 | ||
AF156934.1 | - | 12, 13 | 1737 | |||
AF203445.1 | - | 12, 13 | 1684 | |||
AF203446.1 | - | 12, 13 | 1072 | |||
AF247674.1 | - | 13, 14 | 1468 | |||
AK076282.1 | - | 2210 | ||||
AK145556.1 | - | 12, 13 | 1744 | |||
AK160291.1 | - | 12, 13 | 1746 | |||
AK183147.1 | - | 4, 5 | 397 | |||
BC003252.1 | - | 12, 13 | 1740 | 3500413 | ||
BC055071.1 | - | 4159 | ||||
BC066184.1 | - | 12, 13 | 1719 | 30457053 | ||
BC068314.1 | - | 12, 13 | 1719 | 30461783 |