Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tank | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164071.1 | NP_001157543.1 | 8 | 1264 | ||
NM_001164072.1 | NP_001157544.1 | 7 | 1056 | |||
NM_011529.2 | NP_035659.1 | 7 | 1055 | |||
GenBank mRNA | AK031559.1 | - | 7 | 1018 | ||
AK088817.1 | - | 7 | 1002 | |||
AK147727.1 | - | 8 | 1210 | |||
AK165420.1 | - | 7 | 1012 | |||
AK165994.1 | - | 9 | 1073 | |||
BC026148.1 | - | 7 | 1037 | 3492937 | ||
BC033282.1 | - | 8 | 1112 | |||
BC052685.1 | - | 7 | 1693 | |||
U51907.1 | - | 7 | 994 | |||
U59864.1 | - | 7 | 1068 |