Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK153890.1 | - | 9 | 2038 | ||
BC013802.1 | - | 9 | 1445 | 3257566 | ||
BC024897.1 | - | 9 | 1557 | 3257734 | ||
BC031584.1 | - | 5 | 2169 | |||
KJ650227.1 | - | 9 | 1710 | |||
M55637.1 | - | 9 | 1426 | |||
U60018.1 | - | 9 | 1710 | |||
U60020.1 | - | 9 | 1710 | |||
U60021.1 | - | 9 | 1710 | |||
U60023.1 | - | 9 | 1710 | |||
X59615.1 | - | 9 | 1269 |