Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013684.3 | NP_038712.3 | 7 | 1054 | ||
GenBank mRNA | AK085037.1 | - | 7 | 1055 | ||
BC012685.1 | - | 9 | 1290 | 3498542 | ||
BC016476.1 | - | 6 | 864 | 3584341 | ||
BC050136.1 | - | 7 | 1005 | 30020603 | ||
D01034.1 | - | 6 | 871 | |||
DQ906043.1 | - | 52 | ||||
U63933.1 | - | 8 | 1060 |