Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013684.3 | NP_038712.3 | 3, 4 | 670 | ||
GenBank mRNA | AK085037.1 | - | 3, 4 | 671 | ||
BC012685.1 | - | 5, 6 | 906 | 3498542 | ||
BC016476.1 | - | 3, 4 | 648 | 3584341 | ||
D01034.1 | - | 2, 3 | 487 | |||
U63933.1 | - | 4, 5 | 676 |