Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbrg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025289.3 | NP_079565.2 | 8 | 1226 | ||
GenBank mRNA | AK003270.1 | - | 8 | 1152 | ||
AK010925.1 | - | 8 | 1176 | |||
AK011268.1 | - | 8 | 1151 | |||
AK131158.1 | - | 5 | 3436 | |||
AK150549.1 | - | 8 | 1153 | |||
AK151075.1 | - | 8 | 1104 | |||
AK151209.1 | - | 8 | 1120 | |||
AK151457.1 | - | 8 | 1138 | |||
AK166547.1 | - | 8 | 1148 | |||
BC021331.1 | - | 8 | 1237 | 5003215 | ||
BC040813.1 | - | 8 | 1139 | 4223194 | ||
BC043022.1 | - | 8 | 1141 | 5688749 | ||
BC054057.1 | - | 7 | 1407 | |||
BC065795.1 | - | 8 | 1139 | 1529903 | ||
DQ144541.1 | - | 8 | 1058 |