Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbrg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025289.3 | NP_079565.2 | 6 | 953 | ||
GenBank mRNA | AK003270.1 | - | 6 | 879 | ||
AK010925.1 | - | 6 | 903 | |||
AK011268.1 | - | 6 | 878 | |||
AK131158.1 | - | 4 | 2388 | |||
AK150549.1 | - | 6 | 880 | |||
AK151075.1 | - | 6 | 831 | |||
AK151209.1 | - | 6 | 847 | |||
AK151457.1 | - | 6 | 865 | |||
AK166547.1 | - | 6 | 875 | |||
AK205215.1 | - | 2 | 78 | |||
BC021331.1 | - | 6 | 964 | 5003215 | ||
BC040813.1 | - | 6 | 866 | 4223194 | ||
BC043022.1 | - | 6 | 868 | 5688749 | ||
BC054057.1 | - | 5 | 1134 | |||
BC065795.1 | - | 6 | 866 | 1529903 | ||
DQ144541.1 | - | 6 | 785 |