Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285472.1 | NP_001272401.1 | 5 | 748 | ||
NM_001285476.1 | NP_001272405.1 | 3 | 672 | |||
NM_011532.2 | NP_035662.1 | 3 | 676 | |||
GenBank mRNA | AF326960.2 | - | 3 | 676 | ||
AF349658.1 | - | 5 | 748 | |||
BC139474.1 | - | 3 | 676 | 8862496 | ||
BC145636.1 | - | 3 | 672 | |||
U15565.1 | - | 1 | 73 | |||
U57327.1 | - | 3 | 298 |