Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011540.2 | NP_035670.2 | 2 | 639 | ||
GenBank mRNA | AJ223854.1 | - | 2 | 639 | ||
AK010167.1 | - | 2 | 627 | |||
BC027631.1 | - | 2 | 611 | 1314541 | ||
BC032992.1 | - | 2 | 611 | |||
Y15845.1 | - | 2 | 629 |