Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcf20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114140.1 | NP_001107612.1 | 2 | 1545 | ||
NM_013836.3 | NP_038864.3 | 2 | 1545 | |||
GenBank mRNA | AK122239.1 | - | 1 | 574 | ||
AY007594.1 | - | 2 | 1545 | |||
BC138038.1 | - | 2 | 1518 | 8861057 | ||
BC171946.1 | - | 2 | 1518 |