Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083967.1 | NP_001077436.1 | 17 | 1773 | ||
NM_013685.2 | NP_038713.1 | 17 | 2088 | |||
GenBank mRNA | AK014343.1 | - | 10 | 1157 | ||
AK028748.1 | - | 17 | 1699 | |||
AK081012.1 | - | 17 | 1725 | |||
AK133885.1 | - | 18 | 2500 | |||
AK151433.1 | - | 10 | 1059 | |||
BC014293.1 | - | 10 | 1110 | 4014231 | ||
BC043050.1 | - | 16 | 1548 | 5708370 | ||
S75870.1 | - | 17 | 1660 | |||
U16321.1 | - | 11 | 1147 | |||
U16322.1 | - | 18 | 2060 | |||
X91753.1 | - | 17 | 1712 |