Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130458.1 | NP_001123930.1 | 8 | 1239 | ||
NM_001130459.1 | NP_001123931.1 | 8 | 1208 | |||
NM_015749.3 | NP_056564.1 | 8 | 1242 | |||
GenBank mRNA | AF090686.1 | - | 7 | 1169 | ||
AK133707.1 | - | 8 | 1204 | |||
AK143761.1 | - | 8 | 1210 | |||
AK147176.1 | - | 8 | 1213 | |||
AK161618.1 | - | 8 | 1209 | |||
AK170401.1 | - | 8 | 1228 | |||
AK185167.1 | - | 2 | 160 | |||
AK186678.1 | - | 2 | 160 | |||
AK206141.1 | - | 2 | 258 | |||
AK216881.1 | - | 2 | 257 | |||
BC003720.1 | - | 7 | 1115 | 3601531 |