Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcte1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013688.2 | NP_038716.2 | 2, 3 | 514 | ||
GenBank mRNA | BC145741.1 | - | 1, 2 | 321 | 40131044 | |
CX237405.1 | - | 2, 3 | 407 | |||
M28821.1 | - | 1, 2 | 321 |