Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FKBP14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017946.3 | NP_060416.1 | 1, 2 | 496 | ||
NR_046478.1 | 1, 2 | 496 | ||||
GenBank mRNA | AK000738.1 | - | 1, 2 | 348 | ||
AY358643.1 | - | 1, 2 | 376 | |||
BC005206.1 | - | 1, 2 | 379 | 4042173 | ||
DA166879.1 | - | 1, 2 | 497 | |||
DB498479.1 | - | 1, 2 | 116 | |||
JQ797475.1 | - | 1, 2 | 203 |