Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tead1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166584.1 | NP_001160056.1 | 5 | 831 | ||
NM_001166585.1 | NP_001160057.1 | 3 | 361 | |||
NM_009346.3 | NP_033372.1 | 4 | 768 | |||
GenBank mRNA | AK035595.1 | - | 1 | 2446 | ||
AK140307.1 | - | 5 | 832 | |||
AK148375.1 | - | 4 | 734 | |||
BC052021.1 | - | 3 | 302 | |||
BC060138.1 | - | 3 | 302 | 6410573 | ||
L06865.1 | - | 3 | 307 | |||
L13853.1 | - | 4 | 495 | |||
S74227.1 | - | 4 | 768 |