Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tead4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080979.1 | NP_001074448.2 | 10 | 1562 | ||
NM_011567.2 | NP_035697.3 | 11 | 1691 | |||
GenBank mRNA | AB221623.1 | - | 9 | 992 | ||
AK133915.1 | - | 11 | 1353 | |||
BC130257.1 | - | 9 | 997 | 40142595 | ||
D87965.1 | - | 10 | 1562 | |||
D87966.1 | - | 11 | 1686 | |||
L26343.1 | - | 11 | 1231 | |||
L26344.1 | - | 1176 | ||||
U51743.1 | - | 9 | 977 | |||
X94441.1 | - | 10 | 1122 |